96-Well Clear Flat Bottom Sulfhydryl-BIND™ Microplate
96-Well Clear Flat Bottom Sulfhydryl-BIND™ Microplate
Actual product may vary
Fisherbrand
96-Well Clear Flat Bottom Sulfhydryl-BIND™ Microplate
For assays requiring site-directed orientation of a biomolecule, especially antibodies
Have Questions?
Catalog NumberColor
FKS10499321Clear
Catalog number FKS10499321
Price (SGD)
2,810.00
PK
Add to cart
Color:
Clear
Price (SGD)
2,810.00
PK
Add to cart
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
FormatStandard
LidWithout Lid
Array8 x 12
PackagingIndividually Wrapped
Binding TypeSulfhydryl-BIND
ColorClear
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working75 to 200 μL
SterilityNon-sterile
Surface TreatmentSulfhydryl
Well ShapeFlat Bottom (F-well)
For Use With (Application)Ideal for assays requiring site directed orientation of a biomolecule, especially antibodies
No. per Case50/Cs.
No. per Pack1/Pk.
Quantity50/Cs.
Unit SizePK