Orion™ ROSS™ PerpHecT™ Micro Glass Bodied Combination pH Electrode
Orion™ ROSS™ PerpHecT™ Micro Glass Bodied Combination pH Electrode
Actual product may vary
Orion™ ROSS™ PerpHecT™ Micro Glass Bodied Combination pH Electrode
Orion™ ROSS™ PerpHecT™ Micro Glass Bodied Combination pH Electrode
Orion™ ROSS™ PerpHecT™ Micro Glass Bodied Combination pH Electrode
Orion™ ROSS™ PerpHecT™ Micro Glass Bodied Combination pH Electrode
Fisherbrand
Orion™ ROSS™ PerpHecT™ Micro Glass Bodied Combination pH Electrode
Measure small volumes down to 15 μL in 384 well plates with micro pH electrodes.
Have Questions?
Catalog NumberConnector Types
FKS10495933BNC (Waterproof)
Catalog number FKS10495933
Price (SGD)
2,979.00
EA
Add to cart
Connector Types:
BNC (Waterproof)
Price (SGD)
2,979.00
EA
Add to cart
Thermo Scientific™ Orion™ ROSS PerpHecT™ Micro Combination Electrodes are best suited for use in biological samples, where they are much more versatile than silver chloride electrodes. When used in conjunction with our Orion LogR technology, the glass bulb of the electrode will measure both pH and temperature at the same location, which allows for extremely accurate measurement in small volumes.
Orion™ ROSS Electrodes provide rapid accuracy and temperature response which goes hand in hand with the excellent stability of this reference system. Compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.
Solutions required 
Electrode fill solution
810007     ROSS pH electrode fill solution, 5 x 60 mL bottles
Electrode storage solution
810001     ROSS pH electrode storage solution, 475 mL bottle
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
810199     All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH
                 electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
                 storage bottle
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with 
810007     ROSS pH electrode fill solution 60 mL 1 bottle
Warranty period
12 months from date of purchase
Specifications
DescriptionCombination pH Micro Electrode
Connector TypesBNC (Waterproof)
JunctionGlass Double
MaterialGlass
Temperature (Metric) Operating0°C to 100°C
DriftLess than 0.002 pH units/day
ElectrodeGlass double junction; Refillable
Min. Immersion4.5mm
Storage RequirementsTo ensure a quick response and free-flowing liquid junction the sensing element and reference junction must not be allowed to dry out. Short-term Storage (up to one week): Soak electrode in ROSS pH Electrode Storage Solution, Orion 810001 or use pH Electrode Storage Solution, Orion 910001. To minimize the chance of breakage the 8220BNWP should be attached to an electrode stand and suspended in a beaker that contains pH electrode storage solution. The electrode should not touch the sides or bottom of the beaker. Long-term Storage (more than one week): The reference chamber should be filled and the filling hole securely covered. Cover the sensing element and reference Junction with the protective cap/sleeve containing a few drops of storage solution. Before returning the electrode to use, prepare it as a new electrode.
Length (Metric)155 mm
Length (Metric) Cable1 m
Diameter (Metric) Cap3 mm
Unit SizeEA