Varitip P Tips
Varitip P Tips
Actual product may vary
Fisherbrand
Varitip P Tips
Use these tips to meet a variety of needs for different lab applications. Eppendorf™ Varitip P Tips are designed for aspirating 1 to 10mL from vessels such as beakers.
Have Questions?
Catalog NumberVolume (Metric)
FKS104938911 to 10 mL
Catalog number FKS10493891
Price (SGD)
750.00
PK
Add to cart
Volume (Metric):
1 to 10 mL
Price (SGD)
750.00
PK
Add to cart
Use these tips to meet a variety of needs for different lab applications. Eppendorf™ Varitip P Tips are designed for aspirating 1 to 10mL from vessels such as beakers.
  • A continuously adjustable pipettor for precise pipetting of volumes up to 10mL in 10μL increments
  • Even liquids with a high vapor pressure or viscosity can be pipetted with ease
  • System consists of a dispensing part and the Maxitip air displacement principle
  • No carry over as Maxitip exchanged
  • Valve for Maxitip ensures drip free dispensing of liquids with a high vapor pressure
  • Graduated Maxitip G for compensation of nominal value of Varipette™
Specifications
Volume (Metric)1 to 10 mL
FilteredNon-filtered
For Use With (Application)Aspirating 2.5mL to 10mL from tall, narrow neck vessels
Tip StyleUniversal Fit