DURAN™ Filtering Flask, with glass hose connection, bottle shape
DURAN™ Filtering Flask, with glass hose connection, bottle shape
Actual product may vary
Fisherbrand
DURAN™ Filtering Flask, with glass hose connection, bottle shape
The filtering flask is a thick-walled vessel which is used as a collecting vessel for the filtrate during filtration.
Have Questions?
Catalog NumberCapacity (Metric)
FKS104933215000 mL
Catalog number FKS10493321
Price (SGD)
1,053.00
EA
Add to cart
Capacity (Metric):
5000 mL
Price (SGD)
1,053.00
EA
Add to cart
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Buechner funnels are cylinders with an integrated large-pored sieve plate. Without additional filter paper, only coarse filtration is possible. Buechner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
  • Provision of filtration flasks with a socket has not only made work in preparation and analytical laboratories easier and simpler, but has also reduced the risk of accidents
  • Vacuum resistant due to the heavy wall thickness
  • Fired-on, highly durable white print
  • bottle shape
Specifications
Diameter (Metric) Base185 mm
Capacity (Metric)5000 mL
MaterialBorosilicate Glass (3.3)
For Use With (Application)Separations by vacuum filtration; Filtration at reduced pressure; Filtrieren; Vacuum filtration; Safe working under vacuum
No. per Pack1/Pk.
AutoclavableAutoclavable
Closure Size11 mm fitting
Unit SizeEA