Industrial Fluid Testing Refractometers
Industrial Fluid Testing Refractometers
Actual product may vary
Fisherbrand
Industrial Fluid Testing Refractometers
Designed specifically for checking ratio or concentration of water-soluble industrial fluids
Have Questions?
Catalog number FKS10491825
Price (SGD)
1,204.00
PK
Add to cart
Price (SGD)
1,204.00
PK
Add to cart
Portable testers are ideal for applications including metalworking coolants and lubricants, heat-treating fluids, hydraulic fluids, firefighting foam and plating baths.
  • Automatic temperature compensation provides accurate readings without correction tables or temperature measurement
  • Temperature-compensation range, 15° to 38°C (59° to 100°F)
  • Large, easy-to-read scales and depth of field
  • Portable: no batteries or power cord required
  • Rugged, chemical-resistant housing
IFT10HP Model
  • High Precision optical instrument with improved 0-10 scale 0.1 scale increments for testing low concentration synthetic cutting and grinding fluids and lubricants
  • 10X focusable eyepiece
  • Coated antireflection glass optical components improve illumination and shadowline contrast
IFT40 Rhino Model
  • Sealed housing and eyepiece; IP67 rated water resistant and dustproof
  • Black polycarbonate housing reduces stray light
  • 0-30 arbitrary scale is divided into 0.5 increments; accuracy to ±0.25%
  • Eyepiece with large depth of focus
  • Built for production floor use; impact resistant to withstand repeated drops from three feet
Specifications
Scale Range0 to 30 Brix
Refractive Index1.33 to 1.38
CalibrationDistilled Water
Accuracy±0.2
Increments0.5%
For Use With (Application)Ideal for applications including Metalworking coolants and lubricants, heat treating fluids, hydraulic fluids, firefighting Foam and plating baths
IncludesDurable Plastic carrying case, sample application pipets, calibration adjustment wrench
Temperature Compensation15°C to 38°C
Unit SizePK