Fuego SCS Basic Gas Burner
Fuego SCS Basic Gas Burner
Actual product may vary
Fisherbrand
Fuego SCS Basic Gas Burner
Safe, streamlined, versatile and energy-efficient laboratory gas burner. Wld Tec™ Fuego SCS pro Gas Burner is fabricated of stainless steel and heat-resistant glass.
Have Questions?
Catalog number FKS10485173
Price (SGD)
2,766.00
PK
Add to cart
Price (SGD)
2,766.00
PK
Add to cart
  • Button or foot pedal operation
  • Gas inlet filter
  • DVGW approved
  • Modern design
    • Low casing promotes working comfort
    • Reduced air distrubance
    • Removable burner head for cleaning
    • Integrated drain
  • Universal
    • Accepts stationary natural gas and propane / butane gas supplies, cartridge gas, or gas cylinders
  • Safe
    • Overheating protection
    • Continuous burner head control
    • Burner head clogging detection
    • Residual heat display
    • Environmentally conscious
    • Energy efficiency in accordance with the most recent standards
  • Take-back guarantee
    • Older WLD-TEC burners will be taken back and disposed of in an environmentally compatible manner
  • Robust
    • Stainless steel construction extends to the controls and gas connection
    • Heat-resistant glass display
Specifications
Product TypeBunsen Burner
Gas TypeNatural Gas, Butane, Propane
Length (Metric)103 mm
MaterialStainless Steel
Height (Metric)49 mm
IncludesFoot Pedal and Button Function
Max. Temperature (Metric)1300°C (Natural Gas), 1350°C (Liquid Gas)
No. per Pack1 Ea.
Weight (Metric)700 g
Width (Metric)130 mm
Unit SizePK