Check Order Status
Universal pH Indicator Paper
Fisherbrand
Universal pH Indicator Paper
Provided with a color table to determine color change.
Have Questions?
Catalog Number
pH Range
FKS10482341
1 to 14
Catalog number
FKS10482341
Price (SGD)
55.00
PK
Add to cart
pH Range:
1 to 14
Price (SGD)
55.00
PK
Add to cart
Dispenser roll with indicator strips 5m long
pH range from 1 to 14
4/pack
Specifications
pH Range
1 to 14
Format
Dispenser Roll
Includes
Color Table
Quantity
4/Pk.
Length (Metric)
5 m