Transwell™ Multiple Well Plate with Permeable Polyester Membrane Inserts
Transwell™ Multiple Well Plate with Permeable Polyester Membrane Inserts
Actual product may vary
Fisherbrand
Transwell™ Multiple Well Plate with Permeable Polyester Membrane Inserts
Featuring a microscopically transparent polyester membrane that is TC-treated for optimal cell attachment and growth. Corning™ Transwell™ Multiwell Plate with Polyester Membrane Inserts provide better visibility under phase contrast microscopy and allow assessment of cell viability and monolayer formation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeDescriptionCulture AreaDiameter (Metric) Outer
FKS104821810.4 μm12 inserts in a 24-well plate0.33 cm26.5 mm
FIS07-200-1540.4 μm12 inserts in a 24-well plate0.33 cm26.5 mm
FIS07-200-1610.4 μm12 inserts in a 12-well plate1.12 cm212 mm
FIS07-200-1700.4 μm6 inserts in a 6-well plate4.67 cm224 mm
Catalog number FKS10482181
Price (SGD)
2,356.00
PK
Add to cart
Pore Size:
0.4 μm
Description:
12 inserts in a 24-well plate
Culture Area:
0.33 cm2
Diameter (Metric) Outer:
6.5 mm
Price (SGD)
2,356.00
PK
Add to cart
  • Ideal for transport studies, chemotaxis, co-culture, and microbial pathogenesis studies
  • Thin membrane and straight pore structure provides consistent transport and cell migration
  • Self-centering insert prevents media from wicking between the insert and the outer well
  • Plate: Virgin Polystyrene
  • Membrane: PET
Specifications
Description12 inserts in a 24-well plate
Culture Area0.33 cm2
MaterialPET
Volume (Metric) Well0.6 mL
Pore Density4 x 106 pores/cm2
Pore Size0.4 μm
Diameter (Metric) Outer6.5 mm
QuantityPackaged 12 inserts in a 24 well plate, 4 plates per case
No. per Case48 inserts, 4 plates/Case
Surface CoatingTC-Treated
ColorClear
Surface TreatmentTC-Treated
SterilitySterile
No. of Wells24
Unit SizePK