Nalgene™ Wide-Mouth Straight-Sided PPCO Jars with Closure
Nalgene™ Wide-Mouth Straight-Sided PPCO Jars with Closure
Actual product may vary
Fisherbrand
Nalgene™ Wide-Mouth Straight-Sided PPCO Jars with Closure
Store pastes, solids, specimens and lab supplies in these translucent, wide-mouth, straight-sided jars that offer excellent chemical resistance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FKS104731211000 mL120 mm
2118-000260 mL53 mm
2118-0004125 mL70 mm
FIS11-815-10C125 mL70 mm
2118-0008250 mL70 mm
FIS11-815-10D250 mL70 mm
2118-0016500 mL120 mm
FIS11-815-10E500 mL120 mm
2118-00321000 mL120 mm
FIS11-815-10F1000 mL120 mm
Catalog number FKS10473121
Price (SGD)
217.00
PK
Add to cart
Capacity (Metric):
1000 mL
Closure Size:
120 mm
Price (SGD)
217.00
PK
Add to cart
Store pastes, solids, specimens and lab supplies in Thermo Scientific™ Nalgene™ Wide-Mouth Straight-Sided PPCO Jars. These versatile, translucent jars have excellent chemical resistance and can be autoclaved repeatedly. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. Not liquid leakproof.
Features:
  • Wide mouth and straight sides provide full-access and complete sample retrieval.
  • 15, 30 and 60mL jars have natural polypropylene closures.
  • 125mL to 1L jars have white polypropylene closures with recessed tops for stacking, which helps with organizing the fridge or counter top and maximizing storage space
  • 125 and 250mL sizes fit Osterizer™ and other blenders that have Mason Jar threads; recommended for use with Fluid Transfer Closure DS2153-0700.
  • For 500mL and 1L sizes, remove closure and autoclave separately.
  • Polypropylene copolymer jars are not liquid leakproof, not recommended for liquid transport.
Recycling Information
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Specifications
Capacity32 oz., 1000 mL
Capacity (Metric)1000 mL
Capacity (English)32 oz.
ColorTranslucent
Diameter (English)4.4 in.
IncludesPolypropylene Caps
MaterialPPCO
Neck ID (Metric)112 mm
No. per Case24
No. per Pack6
Weight (Metric)0.18 kg
ClarityTranslucent
MouthWide
ShapeRound
Height (Metric)151 mm
Product TypeStraight-sided Jar
Height (English)6.0 in.
Closure Size120 mm
Diameter (Metric)112 mm
Closure TypeCap
Unit SizePK