Specifications
Rocking Angle7°
Load Bearing Capacity (Metric)10 kg
Speed Range5 to 70 rpm
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Temperature Range (Metric)4°C to 40°C
Wattage50 W
Frequency50/60 Hz
Depth (Metric)420 mm
Dimensions (D x W x H) Interior420 x 360 x 160 mm
Height (Metric)160 mm
Length (Metric) Platform355 mm
Width (Metric) Platform355 mm
ModelSSL4
Humidity RangeUp to 80%
Unit SizePK