BLAUBRAND™ Borosilicate Glass Compact Burettes
BLAUBRAND™ Borosilicate Glass Compact Burettes
Actual product may vary
BLAUBRAND™ Borosilicate Glass Compact Burettes
BLAUBRAND™ Borosilicate Glass Compact Burettes
Fisherbrand
BLAUBRAND™ Borosilicate Glass Compact Burettes
Fast to dismantle and easy to clean. BRAND™ Blaubrand™ Borosilicate Glass Compact Burettes are also simple to repair because all individual components replaceable.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1046434250 mL
FKS1039748225 mL
Catalog number FKS10464342
Price (SGD)
336.00
PK
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
336.00
PK
Add to cart
  • Includes removable PTFE stopcock
  • Fast to dismantle and easy to clean.
  • Simple repair - all individual components replaceable.
  • Borosilicate glass construction (Boro 3.3 to DIN EN ISO 385.)
  • Calibrated to deliver (TD, Ex).
  • Schellbach stripe.
Specifications
Capacity (Metric)50 mL
Length (Metric)800 mm
MaterialBorosilicate Glass
Graduations0.1 mL
IncludesRemovable PTFE Stopcock
Stopcock MaterialPTFE
Tolerance±0.05 mL
ClassClass AS
Certifications/ComplianceDIN EN ISO 385, USP
Unit SizePK