Nunc™ Dishes with UpCell™ Surface
Nunc™ Dishes with UpCell™ Surface
Actual product may vary
Fisherbrand
Nunc™ Dishes with UpCell™ Surface
Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with Thermo Scientific™ Nunc™ Dishes with UpCell™ Surface.
Have Questions?
Catalog NumberCulture AreaGridded
FKS104636658.8 sq. cmNon-Gridded
Catalog number FKS10463665
Price (SGD)
3,551.00
PK
Add to cart
Culture Area:
8.8 sq. cm
Gridded:
Non-Gridded
Price (SGD)
3,551.00
PK
Add to cart
Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with the temperature-responsive surface of Thermo Scientific™ Nunc™ UpCell™ Dishes. Simply transfer the culture to room temperature to collect cells in suspension or in the form of cell sheet with the help of a supportive membrane.
Features:
  • The hydrophicility of the UpCell surface changes with the external temperature, allowing cells to detach
  • No need for dissociation enzymes, ensuring intact cell surface proteins
  • No need for physical scraping, resulting in high cell viability
  • 3 x 3mm grid design in the 60mm and 100mm dishes prevents cells from clumping and enables harvesting of single cell suspension
  • Supportive membranes included in the 35mm dishes allow for harvesting of the cell sheets
  • Shelf Life: 3 years from date of manufacturing
Recommended for:
  • Culture passaging
  • Subsequent single-cell analyses (e.g. flow cytometry)
  • 3D tissue modeling for transplantation
  • For research use only
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. For research and single-use only
Specifications
DescriptionUpCell Dish w/ Supportive Membrane
Culture Area8.8 sq. cm
Volume (Metric) Well3 mL (Well)
Volume (Metric) Working3 mL
TypeCell Culture Dish
Diameter (Metric) Outer35 mm
Height (Metric)12 mm
No. per Pack5
No. per Case30
Surface TreatmentUpCell, temperature responsive
SterilitySterile
GriddedNon-Gridded
Well ShapeRound, Flat Bottom (F-well)
Unit SizePK