Wheaton™ Glass 20mL Scintillation Vials and Urea Caps
Wheaton™ Glass 20mL Scintillation Vials and Urea Caps
Actual product may vary
Fisherbrand
Wheaton™ Glass 20mL Scintillation Vials and Urea Caps
Maintains integrity of seal with variety of cap liners. Wheaton Science Products™ Glass 20mL Scintillation Vials and Urea Caps are suitable for strong solvents and high-purity chemicals.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterial (Liner)Closure SizeColor
FKS10458043Polyethylene Cone22-400White
FIS03-340-25NMetal Foil24-400Clear
FIS03-341-25JPolyethylene Cone22-400White
Catalog number FKS10458043
Price (SGD)
1,489.00
PK
Add to cart
Material (Liner):
Polyethylene Cone
Closure Size:
22-400
Color:
White
Price (SGD)
1,489.00
PK
Add to cart
Maintains integrity of seal with variety of cap liners. Wheaton Science Products™ Glass 20mL Scintillation Vials and Urea Caps are suitable for strong solvents and high-purity chemicals.
  • Chemically inert, suitable for strong solvents and high-purity chemicals
  • Variety of cap liners available to maintain integrity of seal
  • Background counts are consistent and low; ultraviolet transmission is high
  • With screw caps attached to vials
Specifications
Material (Liner)Polyethylene Cone
Diameter (English) Outer1.1 in.
ColorWhite
Diameter (Metric) Outer28 mm
MaterialBorosilicate Glass
Volume (Metric)20 mL
For Use With (Application)Offers superior torque retention and excellent sealing characteristics
Height (English)2.4 in.
Packaging TypeCellular Trays
Volume (English)0.67 oz.
Product LineWheaton
Volume (Metric) Total20 mL
Closure MaterialUrea
Closure TypeScrew Cap
Height (Metric)61 mm
Certifications/ComplianceASTM E438, Type I, Class A
TypeScintillation Vial
Closure Size22-400
Unit SizePK