Polypropylene Cutting Board
Polypropylene Cutting Board
Actual product may vary
Fisherbrand
Polypropylene Cutting Board
Made of non-absorbent polypropylene and easy to clean
Have Questions?
Catalog number FKS10452395
Price (SGD)
186.00
EA
Add to cart
Price (SGD)
186.00
EA
Add to cart
  • Good chemical resistance
  • Unbreakable in normal use
  • Autoclavable at 121°C
Specifications
AutoclavableAutoclavable
MaterialPolypropylene
Size300 L x 210 W x 9 Hmm
Dimensions (L x W)300 x 210 mm
Dimensions (L x W x H)300 x 210 x 9 mm
Length (Metric)300 mm
Width (English)8.26 in.
Width (Metric)210 mm
Height (Metric)9 mm
Unit SizeEA