HandyStep™ S Repetitive Pipet with Rack Mount
HandyStep™ S Repetitive Pipet with Rack Mount
Actual product may vary
Fisherbrand
HandyStep™ S Repetitive Pipet with Rack Mount
The HandyStep™ S Repetitive Pipet offers the highest-precision dispensing of liquid media with high viscosity, density, or vapor pressure, making it ideal for routine or research applications in diagnostics, molecular biology, and environmental analysis.
Have Questions?
Catalog NumberVolume (Metric)
FKS104502902 uL to 5 mL
Catalog number FKS10450290
Price (SGD)
1,309.00
EA
Add to cart
Volume (Metric):
2 uL to 5 mL
Price (SGD)
1,309.00
EA
Add to cart
Ideal for routine or research applications in diagnostics, molecular biology, and environmental analysis, the HandyStep™ S and PD-Tips II offer the highest-precision dispensing of liquid media with high viscosity, density, or vapor pressure. The direct displacement mechanism ensures contamination-free operation, as no aerosols are formed.
  • Perfect for filling individual wells in microtiter plates
  • Suitable for routine dispensing
  • Provides up to 49 repetitive dispensings from a single aspiration
  • Ergonomic shape contoured for comfort and ease of operation
  • Rugged, multifunction lock / filling lever for trouble-free operation
  • Integrated volume chart
  • Nine settings in conjunction with ten PD-Tip II sizes provide 59 different volumes
  • Dispensing lever for smooth, consistent dispensing
  • Delivers 2 μL to 5 mL
  • Contact-free tip ejector
  • Shelf / rack mountable for convenient storage
  • Increased chemical resistance through innovative plastic materials
  • Easy tip mounting - PD-Tip is simply inserted from below
  • Weighs only 108 g
  • Suitable for use with BRAND PD-Tips II, Encode™ tips, REPET-TIPS™, Combitips™, Combitips™ plus and other compatible dispenser tips
  • CE-IVD compliant
Specifications
Compatible TipsPD-Tip II, Dispenser Tips
Adjustable Tip SpacingNo
AutoclavableNot Autoclavable
ColorNeutral
DisposableReusable
For Use WithPD-Tip II
MaterialPolymer, Metal
ModelHandyStep S Mechanical
Number of Channels1
ProgrammabilityNo
SterilityNon-sterile
TypePositive Displacement Repetitive Pipette
Volume (Metric)2 uL to 5 mL
IncludesShelf/rack mount, one each 0.1 mL, 1 mL, 10 mL PD-Tip II syringe tip, operating manual, certificate of performance
Quantity1 Each
Warranty2 Years
Unit SizeEA