Millivac™ Mini Pump
Millivac™ Mini Pump
Actual product may vary
Fisherbrand
Millivac™ Mini Pump
Compact, maintenance-free pumps that provide a consistent source of vacuum for filtration and other laboratory applications
Have Questions?
Catalog NumberElectrical Requirements
FKS10450031230 V 50 Hz
Catalog number FKS10450031
Price (SGD)
5,532.00
EA
Add to cart
Electrical Requirements:
230 V 50 Hz
Price (SGD)
5,532.00
EA
Add to cart
Compact, maintenance-free pumps that provide a consistent source of vacuum for filtration and other laboratory applications
  • A patented diaphragm design has reduced the footprint of the pump compared to conventional models
  • Use Millivac Mini Pump to filter water and aqueous solutions
  • No lubrication required
  • Not explosion-proof
  • 4mm ID tubing
  • Dimensions: 22.6 x 9 x 14.1cm, L x W x H
Specifications
Shipping Weight (English)4.1 lb.
Shipping Weight (Metric)1.9 kg
IncludesHose connector
Electrical Requirements230 V 50 Hz
MaterialRyton (Pump Head), EPDM (Membrane), Fluoroelastomer (Valves)
Weight (English)4.1 lb.
Weight (Metric)1.9 kg
Flow Rate6 L/min.
Frequency50 Hz
Overall Height (English)5.5 in.
Overall Height (Metric)14.1 cm
Overall Length (English)8.8 in.
Overall Length (Metric)22.6 cm
Pump TypeMini Pump
ApplicationsFiltration