Nunc™ Petri Dishes
Nunc™ Petri Dishes
Actual product may vary
Nunc™ Petri Dishes
Nunc™ Petri Dishes
Nunc™ Petri Dishes
Fisherbrand
Nunc™ Petri Dishes
Prepare samples in hospital environments, food science or the pharmaceutical industry with these petri dishes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FKS10444912100 mm
249964140 mm
FIS08-757-100140 mm
FIS08-757-099100 mm
Catalog number FKS10444912
Price (SGD)
692.00
PK
Add to cart
Diameter (Metric):
100 mm
Price (SGD)
692.00
PK
Add to cart
Prepare samples in hospital environments, food science or the pharmaceutical industry with Thermo Scientific™ Nunc™ Petri Dishes. These dishes are available with a molded grid and perform well in automatic dispensers due to complete flatness and uniform height.
  • For the culturing of bacteria, fungi and other microorganisms
  • Constructed of high-quality and optically clear polystyrene, with lid
  • Fitted with top and bottom stacking rings for easier handing and stable stacking
  • Vented for gaseous exchange
  • Sterilized by gamma irradiation
Specifications
Bottom ShapeFlat
Culture Area58 sq. cm
MaterialPolystyrene
ShapeRound
SterilitySterile
Surface TreatmentNunclon Delta
TypePetri Dish
Well Count1
No. per Case320
No. per Pack20
Height (Metric)15 mm
Height (English)0.60 in.
Diameter (Metric)100 mm
Unit SizePK