500μL Polypropylene 96-Well Deep Well Plates
500μL Polypropylene 96-Well Deep Well Plates
Actual product may vary
Fisherbrand
500μL Polypropylene 96-Well Deep Well Plates
Combine a new well design with a sophisticated color coding system for ease of reading and orientation. The well geometry of Eppendorf™ 500μL Polypropylene 96-Well Deep Well Plates allows high speed pipetting without splashing, excellent mixing and high speed centrifugation, which accelerates your manual and automated work processes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorQuantity
FKS10443692Green40 Plates
FKS10185692Blue40 Plates
Catalog number FKS10443692
Price (SGD)
2,227.00
PK
Add to cart
Color:
Green
Quantity:
40 Plates
Price (SGD)
2,227.00
PK
Add to cart
  • Fulfill all current SBS, IVD, EN ISO 9001/2/3 and EN 13485 guidelines.
  • Recovermax - concaved round bottom well shape with smoothed edges
  • Optitrack - color framed plates with alphanumeric labelling
  • High chemical resistance
  • G-safe - highest centrifugation stability
  • LoBind - free of surface coating e.g., silicone
  • Free from DNA, DNase, RNase and PCR inhibitor (PCR clean)
  • LoBind Protein - minimum protein loss (less than 3% BSA 1μg/mL)
  • LoBind DNA - less than 1% loss of DNA/RNA at low concentrations and critical conditions (high salt, short DNA fragments)
  • Autoclavable
  • Optionally available with barcode
Specifications
FormatStandard
Array8 x 12
ColorGreen
MaterialPolypropylene
No. of Wells96
Total Volume (Metric) Well500 μL
Quantity40 Plates
Unit SizePK