Porcelain Evaporating Dishes
Porcelain Evaporating Dishes
Actual product may vary
Fisherbrand
Porcelain Evaporating Dishes
Designed with glazed laboratory porcelain to provide excellent thermal and chemical resistance. Haldenwanger™ Porcelain Evaporating Dishes provide a solid basin for evaporating excessive solvents to produce a concentrated solution or solid precipitate.
Have Questions?
Catalog NumberDiameter (Metric)Height (Metric)Bottom Shape
FKS1044300180 mm32 mmRound
Catalog number FKS10443001
Price (SGD)
48.00
EA
Add to cart
Diameter (Metric):
80 mm
Height (Metric):
32 mm
Bottom Shape:
Round
Price (SGD)
48.00
EA
Add to cart
  • Available in shallow and semi-shallow form
  • Sizes up to 175mL glazed; 1,000mL and larger are glazed except outside base
Specifications
Bottom ShapeRound
Capacity (Metric)75 mL
MaterialPorcelain
ShapeRound
TypeEvaporating Dish
For Use With (Application)Laboratories
Interior FinishGlazed
Height (Metric)32 mm
Diameter (Metric)80 mm
Unit SizeEA