1x1 image pixel for data collection
BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap
BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap
Actual product may vary
Fisherbrand
BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap
Pre-coated with Collagen I for improved cell attachment and an increased proliferation rate of transfected mammalian cells, primary cells and stem cells. Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap provides highly consistent and biologically functional precoated surfaces to more closely mimic in vivo environments for cell culture applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Culture AreaNo. per CaseNeck Style
FKS1044170170 mL25 cm210/Cs.Canted
CRG356487750 mL175 cm240/Cs.Straight
CRG356485250 mL75 cm250/Cs.Canted
Catalog number FKS10441701
Price (SGD)
602.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Capacity (Metric):
70 mL
Culture Area:
25 cm2
No. per Case:
10/Cs.
Neck Style:
Canted
Price (SGD)
602.00
PK
Add to cart
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.
Applications include:
  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in culture
Source
  • Rat tail tendon
  • Quality Control
    • Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
    • Tested and found negative for bacteria and fungi
    • Collagen I purity >90% by SDS-PAGE
    Storage
  • 4°C to 30°C under dry conditions.
  • Specifications
    BarcodeYes
    Capacity (English)2.36 oz.
    Closure MaterialPhenolic
    Capacity (Metric)70 mL
    For Use With (Application)For adhesion, growth and differentiation of neuronal and transfected cell lines
    Neck StyleCanted
    No. per Case10/Cs.
    No. per Pack10/Pk.
    ShapeRectangular
    Surface CoatingCollagen I
    SterilitySterile
    Bottom ShapeRectangular
    Closure TypeVented
    Culture Area25 cm2
    Unit SizePK
    Documents & Downloads
    Certificates
    0 results displayed, search above for a specific certificate
    Share catalog number, name or link