Packaging Bags with Closure
Packaging Bags with Closure
Actual product may vary
Fisherbrand
Packaging Bags with Closure
Designed with zipper-type closure for storing, collecting, protecting and shipping. Buerkle™ Packaging Bags with Closure are suited for specimens, samples, and sales products.
Have Questions?
Catalog NumberCapacity (Metric)Length (Metric)Width (Metric)
FKS104411511000 mL220 mm160 mm
Catalog number FKS10441151
Price (SGD)
89.00
PK
Add to cart
Capacity (Metric):
1000 mL
Length (Metric):
220 mm
Width (Metric):
160 mm
Price (SGD)
89.00
PK
Add to cart
Bags with a patented zipper closure ensures contents remain dust-free. The film is waterproof and has good tear-resistance.
  • LDPE, crystal-clear
  • 0.05 mm strong film
  • Suitable for use with foodstuff
Specifications
AutoclavableNot Autoclavable
Capacity (English)33.81 oz.
Capacity (Metric)1000 mL
Closure TypeZipper
ColorClear
DisposableSingle-use
For Use With (Application)For Storing, Collecting, Protecting and Shipping of Specimens, Samples and Sales Products and Foodstuffs
MaterialLDPE
SterilityNon-sterile
Thickness (Metric)0.05 mm
Quantity100/Pk.
Thickness (English)0.002 in.
Dimensions (L x W)220 x 160 mm
Length (English)8.66 in.
Length (Metric)220 mm
Width (English)6.30 in.
Width (Metric)160 mm
Product TypeZip-seal Storage Bag
Unit SizePK