SampleSafe™ Seal
SampleSafe™ Seal
Actual product may vary
Fisherbrand
SampleSafe™ Seal
Buerkle™ SampleSafe™ Seal feature a one-way tightening mechanism that offers additional security against manipulation.
Have Questions?
Catalog number FKS10434532
Price (SGD)
423.00
PK
Add to cart
Price (SGD)
423.00
PK
Add to cart
  • Clear identification via numbers in sequence
  • 100 seals (10 strips) per pack
  • Protection against manipulation by stainless steel safety spring
Specifications
ColorRed
For Use With (Application)Retention Samples
IncludesNumers in sequence, stainless steel safety spring
Length (English)10.23 in.
Length (Metric)260 mm
Quantity100/Pk.
Weight (English)1.76 lb.
MaterialPolypropylene
TypeSeal
Unit SizePK