Whatman™ Qualitative Filter Paper: Grade 597½ Pleated Circles
Whatman™ Qualitative Filter Paper: Grade 597½ Pleated Circles
Actual product may vary
Fisherbrand
Whatman™ Qualitative Filter Paper: Grade 597½ Pleated Circles
Pleated to save time and achieve optimal fit in conical filter funnels. Whatman™ Qualitative Filter Paper: Grade 597½ Pleated Circles provide extra surface area to increase total loading capacity.
Have Questions?
Catalog NumberDiameter (Metric)
FKS10433141185 mm
Catalog number FKS10433141
Price (SGD)
310.00
PK
Add to cart
Diameter (Metric):
185 mm
Price (SGD)
310.00
PK
Add to cart
Pleated to save time and achieve optimal fit in conical filter funnels. Whatman™ Qualitative Filter Paper: Grade 597½ Pleated Circles provide extra surface area to increase total loading capacity.
Folded filter is used for many routine analytical applications in various industries. Pleated paper enhances and eases filtering process. 
  • Saves time in repetitive or multiple analyses
  • Decreases overall filtration time; normal slow down due to particulate loading is eliminated
  • Increases total loading capacity
  • Maintains flow rate; contact with funnel is reduced, shape of filter is self-supporting
  • Pleating does not significantly affect technical data; same figures may be used with flat circles
  • Grade 597½ is a medium fast paper with medium to fine particle retention
  • Applications include:
    • Food testing; determination of fat content
    • Removal of carbon dioxide and turbidity from beverages
Specifications
DescriptionGrade 597 ½ Qualitative Filter Paper Folded (Prepleated), 185 mm
FormatPre-pleated Circle
MaterialCellulose
Particle Retention7 μm
Pore Size4 to 7 μm
Filtration SpeedApprox. 70 s
Flow RateMedium Fast
For Use With (Application)A prepleated filter used for a wide variety of analytical routine applications in different industries like food testing (e.g., determination of fat content) or removal of carbon dioxide and turbidity from beverages (e.g., beer analysis).
GradeGrade 597 ½
Quantity100 Pk.
Thickness (Metric)180 μm
TypeQualitative Filter Paper
Diameter (English)7.28 in.
Diameter (Metric)185 mm
Unit SizePK