Check Order Status
pH Buffer Sachets, METTLER TOLEDO™
Element Search
Structure Search
Fisherbrand
pH Buffer Sachets, METTLER TOLEDO™
Buffer solutions in sealed sachets for easy and precise calibration of pH measurement equipment.
Have Questions?
Catalog Number
Quantity
pH
FKS10431721
30 x 20 mL
10.0
Catalog number
FKS10431721
Price (SGD)
385.00
PK
Add to cart
Quantity:
30 x 20 mL
pH:
10.0
Request bulk or custom format
Price (SGD)
385.00
PK
Add to cart
Specifications
Color
Blue/White
Concentration
0.02 M
For Use With (Application)
Calibration
For Use With (Equipment)
pH Meter
pH
10.0
View more
Traceable to NIST primary standards