Azlon™ LPDE Slope Shoulder Wash Bottles
Azlon™ LPDE Slope Shoulder Wash Bottles
Actual product may vary
Fisherbrand
Azlon™ LPDE Slope Shoulder Wash Bottles
Feature closures with precision tipped apertures, ideal for fine liquid jet dispensing. Azlon™ LDPE Slope Shoulder Wash Bottles designed to fit a variety of washing needs. Use these durable plastic bottles for a wide range of liquid storage, media preparation and sampling applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Color
FKS10431625250 mLWhite
FKS10004704500 mLWhite
FKS103093141000 mLWhite
Catalog number FKS10431625
Price (SGD)
76.00
PK
Add to cart
Capacity (Metric):
250 mL
Closure Color:
White
Price (SGD)
76.00
PK
Add to cart
Use these durable plastic bottles for a wide range of liquid storage, media preparation and sampling applications. AzlonTM LDPE Slope Shoulder Wash Bottles designed to fit a variety of washing needs. Closures feature precision tipped apertures, ideal for fine liquid jet dispensing.
  • Slim design enables comfortable handling and enhanced ease of use
  • Fitted with a choice of white or red polypropylene swivel dispensing closures
  • Choice of three sizes
  • Closures feature precision jet-tip aperture for fine control
Specifications
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
ColorTranslucent
Diameter (Metric) Outer Neck54mm
MaterialLDPE
Quantity5/PK
Temperature Range (Metric)–50 to 80°C
Closure ColorWhite
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)152 mm
Product TypeWash Bottle
Closure MaterialPolypropylene
Diameter (Metric)152 mm
Closure TypeCap
Through the Closure DispensingYes
Unit SizePK