Nalgene™ PPCO Centrifuge Bottles with Sealing Closure
Nalgene™ PPCO Centrifuge Bottles with Sealing Closure
Actual product may vary
Nalgene™ PPCO Centrifuge Bottles with Sealing Closure
Nalgene™ PPCO Centrifuge Bottles with Sealing Closure
Nalgene™ PPCO Centrifuge Bottles with Sealing Closure
Fisherbrand
Nalgene™ PPCO Centrifuge Bottles with Sealing Closure
Centrifugalize low- to moderate-speed, large-volume, biological and chemical samples with these PPCO centrifuge bottles, which come with a sealing closure.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNominal Capacity
FKS10430613250 mL
3141-0250250 mL
3141-0500450 mL
FIS05-564-3450 mL
FKS10587951450 mL
Catalog number FKS10430613
Price (SGD)
-
Nominal Capacity:
250 mL
Centrifugalize low- to moderate-speed, large-volume, biological and chemical samples with Thermo Scientific™ Nalgene™ PPCO Centrifuge Bottles, which include a sealing closure for added leakproof† assurance at maximum speeds when spinning hazardous materials. Molded of rigid and robust polypropylene copolymer, these translucent bottles are designed for pelleting applications, such as cell culture harvesting and protein biochemistry, including ammonium sulfate precipitations. Additionally, these PPCO bottles are autoclavable and offer excellent mechanical strength.
Features:
  • For low- to moderate-speed centrifugation of biological and chemical samples
  • Bottle includes polypropylene sealing cap assembly with silicone gasket for added leakproof† assurance at maximum speeds or when spinning hazardous materials
  • For proper performance, bottles must be filled to 80% of total capacity
  • Autoclavable
Requires:
When spinning 250mL bottle in a Sorvall GSA rotor, use of the Nalgene centrifuge bottle adapter (DS3125-0250) is required.
Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) centrifuge tubes are designed to last many spins under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These centrifuge tubes are recyclable in many communities. At end of product life, please wash and recycle your Nalgene PP and PPCO centrifuge tubes if services are available in your community.
Specifications
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
ColorTranslucent
Diameter (Metric) Outer Neck60.7 mm
MaterialPPCO
Length (Metric)133.4 mm
No. per Pack4
Nominal Capacity250 mL
Closure IncludedYes
GraduatedNo
Max. Force27,500 x g
ShapeRound
Height (Metric)132.8 mm
Product TypeCentrifuge Bottle
Width (English)2.40 in.
Length (English)5.23 in.
Height (English)5.23 in.
Closure MaterialPolypropylene, red rubber
Diameter (Metric)61.1 mm
Closure TypeSealing Cap Assembly with Silicone Gasket
Through the Closure DispensingNo