Orion™ Ion Selective Electrode Accessories
Orion™ Ion Selective Electrode Accessories
Actual product may vary
Fisherbrand
Orion™ Ion Selective Electrode Accessories
Use Thermo Scientific™ Orion™ Ion Selective Electrode Accessories for solid state and plastic membrane ion selective electrodes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
FKS10428722Ion Selective ElectrodePolishing Strips
FIS13-641-771Ion Selective ElectrodePolishing Strips
Catalog number FKS10428722
Price (SGD)
500.00
PK
Add to cart
For Use With (Equipment):
Ion Selective Electrode
Type:
Polishing Strips
Price (SGD)
500.00
PK
Add to cart
Use Thermo Scientific™ Orion™ Ion Selective Electrode Accessories for solid state and plastic membrane ion selective electrodes.
Electrodes handles are used with plastic membrane combination or half-cell electrodes. Polishing strips help restore the performance of solid-state, half-cell, or combination ion selective electrodes.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionPolishing Strips for Solid State Ion Selective Electrodes
For Use With (Equipment)Ion Selective Electrode
TypePolishing Strips
Unit SizePK