DURAN™ GLS 80™ Screw Cap, Blue PP, Linerless Lip Seal
DURAN™ GLS 80™ Screw Cap, Blue PP, Linerless Lip Seal
Actual product may vary
DURAN™ GLS 80™ Screw Cap, Blue PP, Linerless Lip Seal
DURAN™ GLS 80™ Screw Cap, Blue PP, Linerless Lip Seal
Fisher Catalogue
DURAN™ GLS 80™ Screw Cap, Blue PP, Linerless Lip Seal
Blue PP GLS 80™ wide-mouth screw cap (plug seal) and drip free, PP pouring ring for GLS 80™ bottles
Have Questions?
Catalog number FKS10424382
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Permits opening and closing of the DURAN™ GLS 80™ bottle with only a three-quarter turn. A matching PP pouring ring is also available, permitting clean, drip-free use.
  • Triple thread permits user friendly opening and closing with only a three-quarter turn
  • Cap offer a linerless, integral lip seal
  • Pouring ring for drip-free pouring and clean, safe working
Specifications
DescriptionScrew cap
For Use With (Application)Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping
MaterialPolypropylene
Quantity10/Pk.
Thread Size (Metric)GLS 80
AutoclavableAutoclavable
Diameter (Metric)87 mm
Unit SizePK