Quickfit™ Borosilicate Glass Light Entrainer Receiver
Quickfit™ Borosilicate Glass Light Entrainer Receiver
Actual product may vary
Fisherbrand
Quickfit™ Borosilicate Glass Light Entrainer Receiver
Determine the water content in a solution with the Dean-Stark pattern apparatus. QuickFit™ borosilicate glass light entrainer receiver.
Have Questions?
Catalog NumberCapacity (Metric)Type
FKS104220252 mLLight Entrainer Receiver
Catalog number FKS10422025
Price (SGD)
422.00
EA
Add to cart
Capacity (Metric):
2 mL
Type:
Light Entrainer Receiver
Price (SGD)
422.00
EA
Add to cart
  • Complies with BS 756
  • Dean and Stark pattern receivers for the determination of the water content in petroleum products
  • For use with solvent entrainers which are lighter than water
  • Borosilicate glass with graduated tube
Specifications
For Use With (Application)For use with solvent entrainers which are lighter than water
TypeLight Entrainer Receiver
MaterialBorosilicate Glass
Capacity2 mL
Certifications/ComplianceBS 756
Capacity (Metric)2 mL
For Use With (Equipment)Light entrainer
SizeCone: 24/29, Socket: 19/26
Unit SizeEA