Azlon™ Acrylic Single-Bore Class B Buret
Azlon™ Acrylic Single-Bore Class B Buret
Actual product may vary
Fisherbrand
Azlon™ Acrylic Single-Bore Class B Buret
Suitable for left or right handed use. The Azlon™ Acrylic Single Bore Class B Buret is great for educational laboratories, especially where breakages often occur.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1042065550 mL
Catalog number FKS10420655
Price (SGD)
221.00
EA
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
221.00
EA
Add to cart
Suitable for left or right handed use. The Azlon™ Acrylic Single Bore Class B Buret is great for educational laboratories, especially where breakages often occur.
  • Acrylic construction for strength and transparency
  • Graduations are screen printed in black to ISO 385 Class B accuracy
  • Suitable for use in educational laboratories especially where breakages often occur
  • Unaffected by most dilute aqueous solutions, however unsuitable for use with organic solvents
  • Turning the stopcock through 90° makes it suitable for left or right handed use
  • Stopcocks can be removed for easy cleaning
Specifications
Capacity (Metric)50 mL
MaterialAcrylic
Graduations0.1 mL
For Use With (Application)Left and Right Handed Use
IncludesPMP/Polyethylene Stopcocks
ScaleBlack
Stopcock MaterialPMP, PTFE
Tolerance±0.1 mL
TypeSingle Bore Buret
ClassClass B
Certifications/ComplianceISO 385
Unit SizeEA