Wash Bottle Carrier
Wash Bottle Carrier
Actual product may vary
Wash Bottle Carrier
Wash Bottle Carrier
Fisherbrand
Wash Bottle Carrier
Transports up to 6 wash bottles. Bel-Art™ Wash Bottle Carrier offers safe, convenient, lightweight transportation that is sturdy and lightweight.
Have Questions?
Catalog NumberHolds
FKS10411935Bottles (250 or 500 mL)
Catalog number FKS10411935
Price (SGD)
260.00
EA
Add to cart
Holds:
Bottles (250 or 500 mL)
Price (SGD)
260.00
EA
Add to cart
  • Secure handle affords both comfort and control
  • Chemical resistant Autoclavable at 121°C (250°F)
  • Holds 250 mL(8 oz.) or 500 mL (16 oz.) bottles
Specifications
Product TypeBottle Rack
Array3 x 2
AutoclavableAutoclavable
ColorBlue
HoldsBottles (250 or 500 mL)
MaterialPolypropylene
Positions6
ShapeRectangular, Rounded Corners
Shelves2 Tiers
StackableNo
SterilityNon-sterile
Dimensions (L x W x H)11.25 x 7.62 x 8.87 in. (28.6 x 19.4 x 22.5cm)
Length (Metric)28.6 cm
Length (English)11.25 in.
Width (Metric)19.4 cm
Width (English)7.62 in.
Height (Metric)22.5 cm
Height (English)8.87 in.
Diameter (Metric) Inner74 mm
Unit SizeEA