Two- and Three-Litre Polycarbonate Erlenmeyer Flasks
Actual product may vary
Fisherbrand
Two- and Three-Litre Polycarbonate Erlenmeyer Flasks
Manufactured from optically clear polycarbonate for optimum visibility and durability. Corning™ Two-and Three-Liter Disposable Polycarbonate Erlenmeyer Flasks are ideal for shaker cultures applications.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Bottom Shape
FKS10410482
2000 mL
Non-baffled
FIS10-126-43
2000 mL
Baffled
FIS10-126-38
3000 mL
Non-baffled
FIS10-126-40
3000 mL
Baffled
4 Options
Catalog number FKS10410482
Price (SGD)
1,726.00
PK
Add to cart
Capacity (Metric):
2000 mL
Bottom Shape:
Non-baffled
Price (SGD)
1,726.00
PK
Add to cart
Fernbach design increases surface area for gas exchange
Made from optically-clear polycarbonate
Ideal for shaker culture applications
Baffled bottoms improve mixing and increase gas exchange
Vent caps supplied in every case of product for applications requiring sterile gas exchange
Sterilized by gamma radiation and nonpyrogenic
RNase- and DNase-free
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifications
Capacity (English)67.6 oz.
Closure ColorOrange
Closure MaterialPolypropylene
DescriptionTop style: Threaded
Capacity (Metric)2000 mL
MaterialPolycarbonate
For Use With (Application)For shaker culture applications