Orion™ Gas-Sensing ISE membranes
Orion™ Gas-Sensing ISE membranes
Actual product may vary
Fisherbrand
Orion™ Gas-Sensing ISE membranes
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
FKS10408532Carbon Dioxide Electrode, 9502BNMP
FIS13-641-929Standard Ammonia Electrode, 9512BNWP, 9512
FKS10292441Standard Ammonia Electrode, 9512BNWP, 9512
Catalog number FKS10408532
Price (SGD)
1,063.00
PK
Add to cart
For Use With (Equipment):
Carbon Dioxide Electrode, 9502BNMP
Price (SGD)
1,063.00
PK
Add to cart
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Fresh membranes provide optimum performance with gas-sensing ion selective electrodes. Ammonia electrodes require different membranes if using standard electrode or high performance ammonia electrode.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionFor Carbon Dioxide Electrode 9502BNMP; Four membranes with O-rings
For Use With (Equipment)Carbon Dioxide Electrode, 9502BNMP
TypeGas-Sensing ISE Membrane
Unit SizePK