Super Over-the-Glass Safety Glasses
Super Over-the-Glass Safety Glasses
Actual product may vary
Fisherbrand
Super Over-the-Glass Safety Glasses
Unique frame design that fits comfortably over most prescription spectacles
Have Questions?
Catalog NumberFrame Color
FKS10405120Blue
Catalog number FKS10405120
Price (SGD)
77.00
EA
Add to cart
Frame Color:
Blue
Price (SGD)
77.00
EA
Add to cart
  • Low cost alternative to safety prescription eyewear
  • Uvex permanent lens coating technology: anti-fog coating on the inside of the lens, scratch resistant on the outside
  • Side arms graduate from hard to soft material, gently grips the side of the head for a secure fit
  • Hi resolution lens for improved optical clarity, specifically for wearing two pairs of specs
  • 100% UV protection
  • Clear lens – suitable for most light environments
  • Clear/UV 2-1.2
  • Uvex supravision NC
  • Blue
  • EN 166 1F KN CE
Specifications
Lens CoatingAnti-fog
Certifications/ComplianceEN 166 1 F KN CE
Lens TintClear
Product TypeSuper OTG Safety Glasses
For Use With (Application)Light Environments
Frame ColorBlue
MaterialPolycarbonate
DisposableReusable
Lens MaterialPolycarbonate
Frame MaterialPlastic
UV ProtectionYes
Frame TypeStandard
Unit SizeEA