Vial Decapper Pliers
Vial Decapper Pliers
Actual product may vary
Fisherbrand
Vial Decapper Pliers
Fisherbrand™ Vial Decapper Pliers are designed to easily remove aluminum crimp tops from vials and bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) Outer
FKS1040114830 mm
FIS13-820-04620 mm
Catalog number FKS10401148
Price (SGD)
560.00
PK
Add to cart
Diameter (Metric) Outer:
30 mm
Price (SGD)
560.00
PK
Add to cart
Fisherbrand™ Vial Decapper Pliers are designed to easily remove aluminum crimp tops from vials and bottles. Available in single- or double-action formats, all pliers are smooth and simple to operate. Durable, high-quality construction ensures consistent, dependable performance.
  • Easily remove aluminum crimp-tops from vials and bottles
  • Smooth and simple to operate
  • Durable, high-quality construction offering consistent, dependable performance
  • Padded-handles allow for maximum grip and comfort to relieve stress tension from hands
  • Made from one-piece, corrosion-resistant ASTM-standard stainless steel
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
  • Overall length: 159 mm (6.25 in.)
Specifications
Diameter (Metric) Outer30 mm
MaterialStainless Steel
For Use With (Application)Vial caps of 30mm size
Certifications/ComplianceISO 9002, cGMP
TypePlier-style Decapper
For Use With (Equipment)30 mm Vial Caps
Unit SizePK