PCR Clean Colorless Safe-Lock Centrifuge Tubes
PCR Clean Colorless Safe-Lock Centrifuge Tubes
Actual product may vary
Fisherbrand
PCR Clean Colorless Safe-Lock Centrifuge Tubes
Protect your samples with these centrifuge tubes. Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes feature a hinged Eppendorf Safe–Lock lid for outstanding protection against unintentional opening during incubation and storage.
Have Questions?
Catalog NumberVolume (Metric)
FKS103980311.5 mL
Catalog number FKS10398031
Price (SGD)
328.00
PK
Add to cart
Volume (Metric):
1.5 mL
Price (SGD)
328.00
PK
Add to cart
Protect your samples with these centrifuge tubes. Eppendorf™ PCR Clean Colorless Safe-Lock Centrifuge Tubes feature a hinged Eppendorf Safe–Lock lid for outstanding protection against unintentional opening during incubation and storage.
  • Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
  • g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
  • Precise lid sealing for minimal evaporation rates during long-term storage
  • Guaranteed functionality from -86 to 100°C
  • Large frosted lid and surface on the side for easy labeling
  • Autoclavable when open (121°C, 20 min)
Specifications
ColorNatural
Volume (Metric)1.5 mL
Quantity1000/Pk.
MaterialPolypropylene
AutoclavableAutoclavable
Closure IncludedYes
Product TypeCentrifuge Tube
Unit SizePK