Versilic™ Silicone Tubing
Versilic™ Silicone Tubing
Actual product may vary
Fisherbrand
Versilic™ Silicone Tubing
Use this tubing for biopharmaceutical applications. Versilic™ Silicone Tubing is biologically inert and does not inhibit cell culture. Stays flexible down to -50°C and maintains its chemical, mechanical and electrical performance up to +200°C.
Have Questions?
Catalog NumberDiameter (Metric) InnerDiameter (Metric) OuterThickness (Metric)Overall Length (Metric)
FKS103978548 mm14 mm14 mm10 m
Catalog number FKS10397854
Price (SGD)
1,071.00
EA
Add to cart
Diameter (Metric) Inner:
8 mm
Diameter (Metric) Outer:
14 mm
Thickness (Metric):
14 mm
Overall Length (Metric):
10 m
Price (SGD)
1,071.00
EA
Add to cart
  • Meets the European Pharmacopoeia requirements for silicone elastomer for closures and tubing, USP Class VI biological tests and ISO 10993 biocompatibility (systemic toxicity, irritation, sensitization, citotoxicity, hemocompatibility and Ames test)
  • Meets FDA and BfR positive lists as well as food grade tests according to EC directives
  • This CE marked tubing is a non-invasive medical device
  • May be used for contact up to 29 days
  • Peroxide-cured silicone
  • Biological neutrality
  • FDA/BfR
  • US and European Pharmacopoeia
  • Temperature resistance -50 to +200°C
Specifications
Certifications/ComplianceFDA (21CFR part 177-2600), EEC/EEG/CEE (directives 85/572, 97/48, 2002/72)
AutoclavableAutoclavable
Diameter (English) Inner0.31 in.
Diameter (Metric) Inner8 mm
MaterialSilicone
Diameter (English) Outer0.55 in.
Diameter (Metric) Outer14 mm
Overall Length (English)32.8 ft.
Overall Length (Metric)10 m
Thickness (English)0.55 in.
Thickness (Metric)14 mm
For Use With (Application)For biopharmaceutical and medical applications
Min. Operating Temperature (English)-58°F
Min. Operating Temperature (Metric)-50°C
ColorTranslucent/Transparent
FlexibilityFlexible
Max. Operating Temperature (English)392°F
Max. Operating Temperature (Metric)200°C
Product TypePeroxide-cured Silicone Tubing
Unit SizeEA