Polystyrene Microplates
Polystyrene Microplates
Actual product may vary
Fisherbrand
Polystyrene Microplates
Constructed of crystal grade virgin polystyrene for optimum visibility. Falcon™ Polystyrene Microplates are available with tissue culture-treated and nontreated surfaces.
Have Questions?
Catalog NumberCulture Area
FKS103968812 cm2
Catalog number FKS10396881
Price (SGD)
524.00
PK
Add to cart
Culture Area:
2 cm2
Price (SGD)
524.00
PK
Add to cart
Available with tissue culture-treated and nontreated surfaces. Made of crystal grade virgin polystyrene for optimum visibility.
  • Choose tissue culture-treated or nontreated surfaces
  • Nontreated growth surface is more hydrophobic than tissue culture surface and is similiar to the surface of a bacteriological Petri dish
  • Sterilized by gamma irradiation
  • Lid design permits stable stacking on benchtop or in incubator
  • All plates (except 08-772-2A and 08-772-16) have patented “labyrinth” lid that reduces evaporation of media from wells, minimizes the risk of contamination
  • Deep skirt and serrated edge permit a firmer grip for plate manipulation
  • Exceptional optical clarity allows for visualization of all cell monolayers
  • Frosted writing ends provide clear writing area for quick identification
  • All plates and lids are suitable for both manual and automated handling
Specifications
Array4 x 6
Closure ColorClear
Closure IncludedClosure Included
Closure MaterialPolystyrene
Bottom ShapeFlat
Capacity (Metric)3.5 mL
ColorClear
Culture Area2 cm2
MaterialPolystyrene
ShapeRectangular
SterilitySterile
Surface TreatmentNon-treated
TypeCell Culture Plate
Volume (Metric) Working1.5 to 2.2 mL
Well Count24
IncludesLid
PackagingIndividually Wrapped
Quantity50/Cs.
Unit SizePK