VACUSAFE Aspiration System
VACUSAFE Aspiration System
Actual product may vary
Fisherbrand
VACUSAFE Aspiration System
Use this aspiration system for the safe disposal of biological liquids. Integra Biosciences™ VACUSAFE Aspiration System is a compact, all-in-one laboratory vacuum pump, providing a safe and efficient way to collect and contain biological liquid waste.
Have Questions?
Catalog number FKS10396852
Price (SGD)
11,211.00
EA
Add to cart
Price (SGD)
11,211.00
EA
Add to cart
  • Enables Biosafety level 1, 2 and 3 labs to comply with the stringent user safety guidelines for handling contaminated solutions
  • Maximizes productivity and minimizing noise and operational inconveniences
  • Equipped with vacuum source protection, a shatterproof bottle and a secure stand that holds the bottle in place
  • Includes integrated vacuum pump, complete with collection bottle, bottle handle, tubing set with filter, mains adapter and VACUBOY hand operator (155 510).
  • 3 liter bottle (safety coated glass)
  • Level detection and lid with quick tubing couplings (PVDF)
  • Self-closing connectors to avoid escape of drops or aerosols
  • All-in-one system includes all necessary components
  • Straightforward operation – simply switch on, set the desired vacuum and start working
  • Integrated pump / valve is switched off automatically when the vacuum is established in the bottle
  • Noise of the integrated pump is well below 65 dB(A)
  • Quick tubing connectors speed up bottle removal
Specifications
Includes2 L Glass bottle, Lid with quick couplings and level detection, Tubing set with filter, Mains adapter: 100-240VAC, 50/60Hz, VACUBOY hand operator with 1-channel SS adapter 40 mm
DescriptionVacuSafe Comfort is designed for aspiration, disposal, or filtration of biological /chemical fluids in a clean and professional way.
Flow Rate8 L/min.
Maximum Pressure-300 to -600 mbar (adjustable)
Unit SizeEA