Protect the vacuum source during aspiration with this membrane filter. Integra Biosciences™ Hydrophobic Membrane Filters are designed for use with VACUSAFE Aspiration Systems.
Have Questions?
Catalog Number
DiameterMetric
FKS10393942
Catalog number FKS10393942
Price (SGD)
106.00
EA
Add to cart
Price (SGD)
106.00
EA
Add to cart
Vacusafe Hydrophobic membrane filter
non-sterile, 0.45μm
Diameter 60mm
For use with VACUSAFE Aspiration Systems
VACUSAFE Aspiration System is a compact, all-in-one laboratory vacuum pump, providing a safe and efficient way to collect and contain biological liquid waste.
Specifications
Filter TypeMembrane Filter
For Use With (Equipment)VACUSAFE Aspiration System