Whatman™ Three-piece Filter Funnel
Whatman™ Three-piece Filter Funnel
Actual product may vary
Fisherbrand
Whatman™ Three-piece Filter Funnel
Features quick and easy filtration. Whatman™ Three-piece Filter Funnels dismantle quickly for the insertion of a new filter. Funnels are available with a choice of three plates and come in several sizes.
Have Questions?
Catalog NumberVolume (Metric)Diameter (Metric) MembraneFiltration Area
FKS10387671115 mL70 mm19.6 cm2
Catalog number FKS10387671
Price (SGD)
1,484.00
EA
Add to cart
Volume (Metric):
115 mL
Diameter (Metric) Membrane:
70 mm
Filtration Area:
19.6 cm2
Price (SGD)
1,484.00
EA
Add to cart
Features quick and easy filtration. Whatman™ Three-piece Filter Funnels dismantle quickly for the insertion of a new filter. Funnels are available with a choice of three plates and come in several sizes.
The increased use of high-efficiency glass microfiber filters in modern laboratories has created a demand for simple and effective filter-holding systems. Whatman 3-piece filter funnels have been designed to complement the range of Whatman fine particle retention, rapid flow rate glass microfiber filters.
  • Three-piece construction; funnel is quickly dismantled for insertion of a new filter
  • Glass sealing flanges are ground flat to ensure a good filter seal
  • Retained solids are deposited within the filter circle
  • Edge clamping prevents peripheral loss and possible passage of solution around, rather than through, the filter circle
  • Parts can be quickly and efficiently cleaned
Choice of Three Plates
  • Acrylic - Supplied as standard; suitable for filtration of most aqueous solutions; maximum working temperature 65°C
  • Polypropylene Plate - Optional extra. Recommended for most acids (except concentrated nitric acid and fuming sulfuric acid) at room temperatures; suitable also for most alcohols, glycols, ethers and ketones; maximum working temperature 100°C
  • PTFE - For virtually all common acids, alkalis and solvents at temperatures up to 100°C; maximum working temperature 200°C
Specifications
MaterialPolypropylene
Filtration Area19.6 cm2
IncludesAcrylic plate
TypeFilter Funnel
Volume (Metric)115 mL
Height (English)6.26 in.
Height (Metric)15.9 cm
Diameter (Metric)70 mm
Diameter (Metric) Membrane70 mm
Unit SizeEA