Cardboard Cryobox Inserts, 136mm
Cardboard Cryobox Inserts, 136mm
Actual product may vary
Cardboard Cryobox Inserts, 136mm
Cardboard Cryobox Inserts, 136mm
Fisher Catalogue
Cardboard Cryobox Inserts, 136mm
Combine these inserts with Cryoboxes for flexible cell storage and sample handling. Fisherbrand™ Cardboard Cryobox Inserts feature ultra-low temperature resistant cardboard with water repellent finish for sample storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (Metric)Array
FKS1038457340mm9 x 9
FKS1035400340mm10 x 10
Catalog number FKS10384573
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Height (Metric):
40mm
Array:
9 x 9
  • Ultra low temperature-resistant cardboard
  • For sample storage
  • Temperature-resistant down to -80°C
  • Water-repellent finish
Specifications
Quantity10 Pack
MaterialCardboard
Product TypeGrid Insert
For Use With (Application)Sample storage
HoldsSample storage tubes
Height (Metric)40mm
Width (Metric)136mm
Length (Metric)136mm
Array9 x 9
Unit SizePK