Orion™ Fluoride Ion Selective Electrodes
Orion™ Fluoride Ion Selective Electrodes
Actual product may vary
Orion™ Fluoride Ion Selective Electrodes
Orion™ Fluoride Ion Selective Electrodes
Orion™ Fluoride Ion Selective Electrodes
Orion™ Fluoride Ion Selective Electrodes
Fisherbrand
Orion™ Fluoride Ion Selective Electrodes
Quickly, simply, accurately and economically measure free fluoride ions in aqueous solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector Types
FKS10374141BNC (Waterproof)
FIS13-642-265BNC (Waterproof)
Catalog number FKS10374141
Price (SGD)
4,272.00
EA
Add to cart
Connector Types:
BNC (Waterproof)
Price (SGD)
4,272.00
EA
Add to cart
Measure free fluoride ions in aqueous solutions quickly, simply, accurately, and economically with Thermo Scientific™ Orion™ Fluoride Electrodes.
The ThermoFisher Orion Fluoride electrode complies with SM 4500-F- C-1997 and ASTM D1179-04 (B) for Drinking water and Wastewater measurements.
Features:
  • 9609BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
  • 9409BN and 9409SC Fluoride Half-Cell Electrode are to be used with the Orion 900100 sure flow single Junction reference electrode
  • 9409SC requires a separate electrode cable with a screw cap connector.
Solutions required:
  • Standards
    • 940906 0.1 M NaF, 475 mL bottle
    • 940907 100 ppm F-, 475 mL bottle
    • 040908 10 ppm F- with TISAB II, 475 mL bottle
    • 040907 2 ppm F- with TISAB II, 475 mL bottle
    • 040906 1 ppm F- with TISAB II, 475 mL bottle
  • ISA
    • 940909 TISAB II, 3.8 L bottle
    • 940999 TISAB II, 4 x 3.8 L bottle
    • 940911 TISAB III concentrate, 475 mL bottle
  • Electrode fill solution
    • 900061 Optimum Results A electrode filling solution 5 x 60 mL bottles
    • 900001 Single junction reference electrode filling solution, 5 x 60 mL bottles
Items included with 9609BNWP only:
  • 900061 Optimum Results A electrode filling solution. 60mL 1 bottle
Warranty period:
  • 12 months from date of purchase
Specifications
Connector TypesBNC (Waterproof)
DescriptionFluoride Ionplus Sure-Flow Solid State Combination
Diameter (English) Body0.51 in.
Diameter (Metric) Body13mm
Filling Solution Type900061 (9609BNWP and 960900); 900001 with 900100 single junction reference electrode (9409BN, 9409SC, and 940900)
JunctionSure-Flow Junction
Length (English)4.33 in.
Length (English) Cable39 in.
Length (Metric)110mm
Length (Metric) Cable1m
MaterialEpoxy
Temperature (English) OperatingContinuous: 32 to 176 °F; Intermittent: 176°F to 212°F
For Use With (Equipment)ISE Meter
Temperature (Metric) Operating0°C to 80°C (Continuous), 80°C to 100°C (Intermittent)
TypeCombination ISE
Measurement Range1 x 10-6 M to saturated 0.02 ppm to saturated
ParameterCombination Ion Selective Electrode
RefillableYes
Unit SizeEA