Gridded Sterile Cellulose Nitrate Membrane Filters: 0.45μm
Gridded Sterile Cellulose Nitrate Membrane Filters: 0.45μm
Actual product may vary
Fisherbrand
Gridded Sterile Cellulose Nitrate Membrane Filters: 0.45μm
Ensure quality control in microbiological filtering. Sartorius Gridded Sterile Cellulose Nitrate Membrane Filters, 0.45μm are single-packed membranes with colored grids for colony counting, particle testing and microscopy. The 0.45μm pore size is made in accordance with ISO 7704. Choose from two diameters and quantities in a range of filter/grid color combinations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)ColorTypeQuantity
FKS1037329147 mmWhiteType 114 Membrane Filter100/Pk.
FIS14-555-59747 mmWhiteType 114 Membrane Filter100/Pk.
FIS14-555-61247 mmWhiteType 114 High Flow Membrane Filter1000/Pk.
Catalog number FKS10373291
Price (SGD)
575.00
PK
Add to cart
Diameter (Metric):
47 mm
Color:
White
Type:
Type 114 Membrane Filter
Quantity:
100/Pk.
Price (SGD)
575.00
PK
Add to cart
Ensure quality control in microbiological filtering. Sartorius Gridded Sterile Cellulose Nitrate Membrane Filters, 0.45μm are single-packed membranes with colored grids for colony counting, particle testing and microscopy. The 0.45μm pore size is made in accordance with ISO 7704. Choose from two diameters and quantities in a range of filter/grid color combinations.
Sterile, individually packaged filters for strict quality control
  • Pre-sterilized via gamma irradiation at 25kGray
  • Ready-to-use to save preparatory time
  • Avoids contamination of remaining filters in opened packs
  • Filter identification and lot number are printed on each individual envelope
  • Manufactured in accordance with ISO 7704, ISO 7899-2, ISO 8199, ISO 9308-1 and EN 12780
  • Each membrane is checked to ensure:
    • It is not damaged in any way
    • It is positioned correctly with no slippage under the edge seal
    • It is free of particles
    • Each envelope has readable lettering
High-quality Cellulose Nitrate membranes
  • Defined particle retention and outstanding, reliable recovery rates for microorganisms
  • Cellulose nitrate filters (mixed cellulose ester) combine high flow rates with optimal colony growth
  • The various-colored grids divide the filtration area into squares and provide the best possible color contrast
    • Enables easy and reliable counting and identification of colonies and particles
    • Does not influence the growth of bacteria and micro-colonies
  • 0.45μm filters are manufactured in accordance with ISO 7704 standards
  • Available in 47 and 50mm (1.9 or 2.0 in.) diameters
Specifications
ColorWhite
DisposableSingle-use
Filter TypeMembrane Filter
FormatDisc
MaterialCellulose Nitrate
Material (Membrane)Cellulose Nitrate
Pore Size0.45 μm
SterilitySterile
Flow RateStandard
For Use With (Application)For colony count; sterility testing; particle count; microscopy
Grid ColorBlack
HydrophobicityHydrophilic
Individually wrappedYes
Membrane TypeGridded
Quantity100/Pk.
TypeType 114 Membrane Filter
Diameter (Metric)47 mm
Unit SizePK