Volac™ Borosilicate Glass Burettes with Glass Key
Volac™ Borosilicate Glass Burettes with Glass Key
Actual product may vary
Fisherbrand
Volac™ Borosilicate Glass Burettes with Glass Key
Featuring a flange at the top for ease of filling. Poulten Graf™ Volac™ Borosilicate Glass Burettes with Glass Key feature bold figures and fine graduation lines.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FKS1037102250 mL0.1 mL
Catalog number FKS10371022
Price (SGD)
266.00
PK
Add to cart
Capacity (Metric):
50 mL
Graduations:
0.1 mL
Price (SGD)
266.00
PK
Add to cart
  • Removable glass jets are inscribed with instrument number for traceability
  • Flow time inscribed on the instrument
  • Certificate specifying accuracy at three points provided
  • Produced according to BS ISO 385
Specifications
ColorBlue
Capacity (Metric)50 mL
Length (Metric)820 mm
MaterialBorosilicate Glass
Graduations0.1 mL
IncludesInterchangeable Glass Key in Glass Barrel
StopcockStraight
Stopcock MaterialGlass
Tolerance±0.05 mL
TypeVolumetric Buret
ClassClass AS
Certifications/ComplianceBS 846, ISO 385
Unit SizePK