Glass Combination pH Electrodes
Glass Combination pH Electrodes
Actual product may vary
Fisherbrand
Glass Combination pH Electrodes
Sensing electrode made with a silver ring with a design that is ideal for use in argentometric titrations.
Have Questions?
Catalog NumberLength (Metric)
FKS10360101120 mm
Catalog number FKS10360101
Price (SGD)
877.00
EA
Add to cart
Length (Metric):
120 mm
Price (SGD)
877.00
EA
Add to cart
  • Recommended for use with pH 20 and pH 21 pH meters
  • For general laboratory use
  • Combination pH Electrode
  • Glass body ORP half-cell electrode with a BNC connector
  • Sensing electrode made with a silver ring and is to be used in conjunction with a reference half-cell electrode
  • Design consideration ideal for use in argentometric titrations
Specifications
Connector TypesBNC
Diameter (Metric)12 mm
TypepH Combination Electrode
For Use With (Application)For laboratory and beer testing
Includes1m Coaxial Cable, BNC Connector
Length (Metric)120 mm
MaterialGlass
pH Range0 to 13
Temperature (Metric) Range20°C to 40°C
Unit SizeEA