1x1 image pixel for data collection
Quebec™ Darkfield Manual Colony Counters
Quebec™ Darkfield Manual Colony Counters
Actual product may vary
Fisherbrand
Quebec™ Darkfield Manual Colony Counters
Have Questions?
Catalog NumberVoltage
FKS10359994220 V
Catalog number FKS10359994
Price (SGD)
7,430.00
EA
Add to cart
Voltage:
220 V
Price (SGD)
7,430.00
EA
Add to cart
The Darkfield Quebec colony counter allows the user to quickly and accurately count bacteria colonies. It is available in two models for digital or manual counting. Both models provide even, glare-free illumination as light is spread uniformly over the entire culture plate.
  • Adjustable dish holder for centering both round dishes and square culture plates
  • Adjustable focusing rod
  • Lens rotates a full 360°
  • Built-in tilt leg
  • Optional 1.5x auxiliary lens fits over standard lens, increasing magnification to 3x
Specifications
Voltage220 V
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link