Tissue Culture Treated Flasks
Actual product may vary
Tissue Culture Treated Flasks
Tissue Culture Treated Flasks
Fisherbrand
Tissue Culture Treated Flasks
Manufactured from optically clear virgin polystyrene for easy viewing of sample or cells. Falcon™ Tissue Culture Treated Flasks features vacuum-gas plasma tissue culture treatment to ensure consistent growth surface.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Culture AreaClosure TypeNeck Style
FKS1035039150 mL25 cm²VentedCanted
FIS10-126-950 mL25 cm²VentedCanted
FIS10-126-1070 mL25 cm2VentedCanted
FIS13-680-65250 mL75 cm2VentedCanted
FIS10-126-8750 mL175 cm2Plug SealStraight
FIS10-126-13750 mL175 cm2VentedStraight
FIS14-826-80800 mL225 cm2VentedCanted
Catalog number FKS10350391
Price (SGD)
391.00
Pack
Capacity (Metric):
50 mL
Culture Area:
25 cm²
Closure Type:
Vented
Neck Style:
Canted
Price (SGD)
391.00
Pack
  • Secure stacking provides stability from accidental tips or spills
  • Nonpyrogenic tested to less than 0.1 EU/ mL
  • Volumetric graduations and writing patch
  • Vented caps incorporate a 0.2 μm hydrophobic membrane for optimal gas exchange
  • Growth area and volume are nominal
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifications
Capacity (English)1.69 oz.
Closure MaterialPolyethylene
Capacity (Metric)50 mL
MaterialPolystyrene
For Use With (Application)For enhanced cell attachment and growth
Neck StyleCanted
No. per Case100/Cs.
No. per Pack20/Pk.
ShapeRectangular
Surface CoatingTC-Treated
SterilitySterile
Bottom ShapeRectangular
Closure TypeVented
Culture Area25 cm²
Unit SizePack