Polystyrene Macro Cuvettes for UV and Visible Wavelengths
Polystyrene Macro Cuvettes for UV and Visible Wavelengths
Actual product may vary
Fisherbrand
Polystyrene Macro Cuvettes for UV and Visible Wavelengths
Use these cuvettes for more constant sample temperatures during photometric measurements. Fisherbrand™ Polystyrene Macro Cuvettes for UV and Visible Wavelengths feature excellent optical transmission range and small wall thickness for increased heat transfer.
Have Questions?
Catalog NumberCapacity (Metric)
FKS103493344 mL
Catalog number FKS10349334
Price (SGD)
176.00
PK
Add to cart
Capacity (Metric):
4 mL
Price (SGD)
176.00
PK
Add to cart
Cuvets with improved photometric properties.
  • Very low variation of extinction values
  • Excellent optical transmission range
Specifications
TypeMacro Cuvette
DisposableReusable
Capacity (Metric)4 mL
MaterialPlastic
Quantity100/Pk.
Wavelength Range220 to 900 nm
Height (Metric)45 mm
Width (Metric)12.5 mm
Unit SizePK