Azlon™ Straight-sided Beakers
Azlon™ Straight-sided Beakers
Actual product may vary
Fisherbrand
Azlon™ Straight-sided Beakers
Manufactured from polypropylene for strength and good chemical resistance. Azlon™ Straight-sided Beakers provide an excellent solution for measuring and dispensing in a laboratory setting and are available in sizes to fit many needs.
Have Questions?
Catalog NumberCapacity (Metric)
FKS103493241000 mL
Catalog number FKS10349324
Price (SGD)
38.00
PK
Add to cart
Capacity (Metric):
1000 mL
Price (SGD)
38.00
PK
Add to cart
Manufactured from polypropylene for strength and good chemical resistance. Azlon™ Straight-sided Beakers provide an excellent solution for measuring and dispensing in a laboratory setting and are available in sizes to fit many needs.
  • Autoclavable to 121deg.C
  • Ideal for general laboratory use - straight stable shape
  • Squat form, rimmed edge, wide spout
  • Graduations are clearly printed in indelible solvent resistant ink
  • Complies with ISO 7056
Specifications
ColorTranslucent
Diameter (Metric) Outer127 mm
Diameter (Metric)Top127 mm
Capacity (Metric)1000 mL
MaterialPolypropylene
For Use With (Application)Ideal for General Laboratory Use
Height (Metric)126 mm
Increments100 mL
Quantity5 Pk.
Temperature (Metric) Max. Operating-20°C to +135°C
TypeBeaker
AutoclavableAutoclavable
GraduatedYes
Certifications/ComplianceISO 7056, FDA
Unit SizePK